Please login to your online account to display your discounted pricing

Trimethylsilyl acetate 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Trimethylsilyl acetate
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C5H12O2Si
Linear Formula CH3COOSi(CH3)3
Formula Weight 132.23
MDL Number MFCD00008695
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC391980250

  SDS
thermo fisher scientific
AC391980250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC391981000

  SDS
thermo fisher scientific
AC391981000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 132.23
Quantity 25g
Color Clear colorless to pale yellow
Density 0.884g/mL
Packaging Glass bottle
Linear Formula CH3COOSi(CH3)3
Assay Percent Range 97%
Chemical Name or Material Trimethylsilyl acetate
MDL Number MFCD00008695
Refractive Index 1.3870 to 1.3890 (20°C, 589nm)
Infrared Spectrum Authentic
CAS 2754-27-0
Molecular Formula C5H12O2Si
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Highly flammable liquid and vapor.