missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trimethylsilyl acetate 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC391981000

 View more versions of this product

Catalog No. AC391981000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥96.0%
132.23
Clear colorless to pale yellow
CH3COOSi(CH3)3
C5H12O2Si
2754-27-0
1.3870 to 1.3890 (20°C, 589nm)
97%
Liquid
100g
0.884g/mL
Authentic
Trimethylsilyl acetate
MFCD00008695
Glass bottle

Chemical Identifiers

97%
CH3COOSi(CH3)3
MFCD00008695
≥96.0%
C5H12O2Si
132.23
Trimethylsilyl acetate

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger