missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Trimethylphosphine, 98%

Manufacturer:  Alfa Aesar™ 3014306

 View more versions of this product

Catalog No. AA3014306



Specifications

Specifications

Trimethylphosphine
−30° (−22°F)
Air & Moisture Sensitive
5g
C3H9P
UN2845
YWWDBCBWQNCYNR-UHFFFAOYSA-N
trimethylphosphane
68983
76.08
99%
0.735g/mL
Pungent
38°C to 40°C
594-09-2
MFCD00008510
trimethylphosphine, trimethyl phosphine, pme3, trimethylphosphorus, phosphine, trimethyl, me3p, ch3 3p, trimethylphospine, trimethylphosphine?, phosphine,trimethyl
CP(C)C
76.079
CHEBI:35890
Liquid, Ampouled Under Argon

Chemical Identifiers

594-09-2
76.079
YWWDBCBWQNCYNR-UHFFFAOYSA-N
68983
trimethylphosphane
C3H9P
MFCD00008510
trimethylphosphine, trimethyl phosphine, pme3, trimethylphosphorus, phosphine, trimethyl, me3p, ch3 3p, trimethylphospine, trimethylphosphine?, phosphine,trimethyl
CHEBI:35890
CP(C)C
Safety and Handling

Safety and Handling

GHS H Statement
H225-H250-H252-H314-H318
Highly flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.

SDS Documents