missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trimethyloxonium Tetrafluoroborate, TCI America™

Manufacturer:  TCI America T150725G

 View more versions of this product

Catalog No. T150725G



Specifications

Specifications

Trimethyloxonium Tetrafluoroborate
25g
C3H9BF4O
1759
CZVZBKHWOFJNCR-UHFFFAOYSA-N
trimethyloxidanium;tetrafluoroborate
2735153
Crystal-Powder at 20°C
Undesignated
420-37-1
MFCD00011798
trimethyloxonium tetrafluoroborate, trimethyloxidanium tetrafluoroborate, tetrafluoroboranuide; trimethyloxidanium, trimethyloxonium fluoborate, acmc-209jn1, ksc236c1n, trimethyloxonium tetrafluroborate, trimethyloxonium tetrafloroborate, trimethyloxonium tetrafluorborate, trimethyloxonium tetraflouroborate
[B-](F)(F)(F)F.C[O+](C)C
147.908
147.91
≥95.0% (T)

Chemical Identifiers

420-37-1
147.908
CZVZBKHWOFJNCR-UHFFFAOYSA-N
2735153
[B-](F)(F)(F)F.C[O+](C)C
C3H9BF4O
MFCD00011798
trimethyloxonium tetrafluoroborate, trimethyloxidanium tetrafluoroborate, tetrafluoroboranuide; trimethyloxidanium, trimethyloxonium fluoborate, acmc-209jn1, ksc236c1n, trimethyloxonium tetrafluroborate, trimethyloxonium tetrafloroborate, trimethyloxonium tetrafluorborate, trimethyloxonium tetraflouroborate
trimethyloxidanium;tetrafluoroborate
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Freezer

Documents