Learn More
Trimethyloxonium tetrafluoroborate, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1517530
Description
Trimethyloxonium tetrafluoroborate act as a avmethylating agent and activates C-X multiple bonds. It is involved in the esterification of polyfunctional carboxylic acids. It acts as a catalyst in the polymerization of cyclic sulfides and ethers. Further, it is used in Beckmann rearrangement of oximes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTrimethyloxonium tetrafluoroborate act as a avmethylating agent and activates C-X multiple bonds. It is involved in the esterification of polyfunctional carboxylic acids. It acts as a catalyst in the polymerization of cyclic sulfides and ethers. Further, it is used in Beckmann rearrangement of oximes.
Solubility
Soluble in nitrobenzene, nitromethane, chloroform, hot acetone, liquid sulfur dioxide. Slightly soluble in dichloromethane. Insoluble in common organic solvents.
Notes
Hygroscopic. Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and bases.
Specifications
Trimethyloxonium Tetrafluoroborate | |
∽190°C (decomposition) | |
C3H9BF4O | |
250 g | |
Moisture sensitive | |
Soluble in nitrobenzene,nitromethane,chloroform,hot acetone,liquid sulfur dioxide. Slightly soluble in dichloromethane. Insoluble in common organic solvents. | |
C[O+](C)C.F[B-](F)(F)F | |
147.91 | |
147.91 |
420-37-1 | |
96% | |
MFCD00011798 | |
3597303 | |
trimethyloxonium tetrafluoroborate, trimethyloxidanium tetrafluoroborate, tetrafluoroboranuide; trimethyloxidanium, trimethyloxonium fluoborate, acmc-209jn1, ksc236c1n, trimethyloxonium tetrafluroborate, trimethyloxonium tetrafloroborate, trimethyloxonium tetrafluorborate, trimethyloxonium tetraflouroborate | |
CZVZBKHWOFJNCR-UHFFFAOYSA-N | |
tetrafluoroboranuide; trimethyloxidanium | |
2735153 |
Chemical Identifiers
420-37-1 | |
147.91 | |
CZVZBKHWOFJNCR-UHFFFAOYSA-N | |
2735153 |
C3H9BF4O | |
MFCD00011798 | |
trimethyloxonium tetrafluoroborate, trimethyloxidanium tetrafluoroborate, tetrafluoroboranuide; trimethyloxidanium, trimethyloxonium fluoborate, acmc-209jn1, ksc236c1n, trimethyloxonium tetrafluroborate, trimethyloxonium tetrafloroborate, trimethyloxonium tetrafluorborate, trimethyloxonium tetraflouroborate | |
C[O+](C)C.F[B-](F)(F)F |
Safety and Handling
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 206-994-4
TSCA : Yes
storageNote1 : Moisture sensitive
Recommended Storage : Store at -20°C; Store under Nitrogen
RUO – Research Use Only