missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trimethylolpropane Tris(thioglycolate), TCI America™

Manufacturer:  TCI America T2633500G

 View more versions of this product

Catalog No. T2633500G



Specifications

Specifications

Trimethylolpropane Tris(thioglycolate)
Yellow
10193-96-1
MFCD00046836
KAJBSGLXSREIHP-UHFFFAOYSA-N
2,2-bis[(2-sulfanylacetyl)oxymethyl]butyl 2-sulfanylacetate
82439
Clear Liquid at 20°C
245°C
500g
C12H20O6S3
1,1,1-Tris(thioglycoloyloxymethyl)propane
CCC(COC(=O)CS)(COC(=O)CS)COC(=O)CS
356.466
356.47
≥80.0% (GC)

Chemical Identifiers

10193-96-1
356.466
KAJBSGLXSREIHP-UHFFFAOYSA-N
82439
CCC(COC(=O)CS)(COC(=O)CS)COC(=O)CS
C12H20O6S3
MFCD00046836
1,1,1-Tris(thioglycoloyloxymethyl)propane
2,2-bis[(2-sulfanylacetyl)oxymethyl]butyl 2-sulfanylacetate
Safety and Handling

Safety and Handling

EINECSNumber : (7)-1074

TSCA : Yes

Documents