Learn More
Trimethylolpropane triacrylate, stabilized, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01644536
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Trimethylolpropane triacrylate | |
15625-89-5 | |
>200°C (1 mmHg) | |
Pungent | |
1.4735 | |
500 g | |
Miscible with water. | |
CCC(COC(=O)C=C)(COC(=O)C=C)COC(=O)C=C | |
296.32 | |
CHEBI:35028 | |
Liquid |
Stabilized | |
1.10 g/mL | |
87°C (188°F) | |
C15H20O6 | |
MFCD00008628 | |
trimethylolpropane triacrylate, tmpta, viscoat 295, nk ester a tmpt, sartomer sr 351, ogumont t 200, saret 351, setalux uv 2241, ccris 92, unii-4b67kgl96s | |
DAKWPKUUDNSNPN-UHFFFAOYSA-N | |
2,2-bis(prop-2-enoyloxymethyl)butyl prop-2-enoate | |
27423 | |
296.31 |
Chemical Identifiers
15625-89-5 | |
296.32 | |
DAKWPKUUDNSNPN-UHFFFAOYSA-N | |
27423 | |
2,2-bis(prop-2-enoyloxymethyl)butyl prop-2-enoate |
C15H20O6 | |
MFCD00008628 | |
trimethylolpropane triacrylate, tmpta, viscoat 295, nk ester a tmpt, sartomer sr 351, ogumont t 200, saret 351, setalux uv 2241, ccris 92, unii-4b67kgl96s | |
CHEBI:35028 | |
CCC(COC(=O)C=C)(COC(=O)C=C)COC(=O)C=C |
Safety and Handling
GHS H Statement
H315-H319-H317-H227
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Combustible liquid.
P210-P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P362-P370+P378q-P501c
H227-H315-H317-H319
EINECSNumber : 239-701-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only