missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Trimethylolpropane triacrylate, stabilized

Manufacturer:  Alfa Aesar™ 1644522

 View more versions of this product

Catalog No. AA1644522



Specifications

Specifications

Trimethylolpropane triacrylate
87° (188°F)
1.4735
100g
15625-89-5
MFCD00008628
DAKWPKUUDNSNPN-UHFFFAOYSA-N
2,2-bis(prop-2-enoyloxymethyl)butyl prop-2-enoate
27423
296.31
1.10g/mL
Pungent
>200°C (1mmHg)
Stabilized
C15H20O6
trimethylolpropane triacrylate, tmpta, viscoat 295, nk ester a tmpt, sartomer sr 351, ogumont t 200, saret 351, setalux uv 2241, ccris 92, unii-4b67kgl96s
CCC(COC(=O)C=C)(COC(=O)C=C)COC(=O)C=C
296.319
CHEBI:35028
Liquid

Chemical Identifiers

15625-89-5
296.319
DAKWPKUUDNSNPN-UHFFFAOYSA-N
27423
2,2-bis(prop-2-enoyloxymethyl)butyl prop-2-enoate
C15H20O6
MFCD00008628
trimethylolpropane triacrylate, tmpta, viscoat 295, nk ester a tmpt, sartomer sr 351, ogumont t 200, saret 351, setalux uv 2241, ccris 92, unii-4b67kgl96s
CHEBI:35028
CCC(COC(=O)C=C)(COC(=O)C=C)COC(=O)C=C
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H317-H227
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents