Please login to your online account to display your discounted pricing

Trimethyleorthoformate, +99%, anhydrous, AcroSeal™, +99%, ACROS Organics™


Quantity 100mL

Manufacturer: thermo fisher scientific  AC429281000

 View more versions of this product

Catalog No. AC429281000

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 106.12
Quantity 100mL
Density 0.9700g/mL
Assay Percent Range 99+%
Linear Formula CH(OCH3)3
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Highly flammable liquid and vapor.
Molecular Formula C4H10O3
Chemical Name or Material Trimethyl orthoformate
CAS 149-73-5
Synonym Methyl orthoformate
Health Hazard 1 Danger

Specifications

Chemical Name or Material Trimethyl orthoformate
Assay Percent Range 99+%
Molecular Formula C4H10O3
Linear Formula CH(OCH3)3
Formula Weight 106.12
Synonym Methyl orthoformate
Accessories

Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Highly flammable liquid and vapor.

SDS & Safety Documents