Please login to your online account to display your discounted pricing

Alfa Aesar™ Trimethylenebis(triphenylphosphonium bromide), 98+%

$186.84 - $458.05

Chemical Identifiers

Chemical Name or Material Trimethylenebis(triphenylphosphonium bromide)
CAS 7333-67-7
Molecular Formula C39H36Br2P2
Formula Weight 726.48
MDL Number MFCD00051885
Beilstein 4281936
Synonym 1,3-Bis(triphenylphosphonio)propane dibromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2516514

  SDS
alfa aesar™
B2516514
25g Each for $186.84

AAB2516522

  SDS
alfa aesar™
B2516522
100g Each for $458.05
Description & Specifications

Specifications

Formula Weight 726.48
Quantity 25g
Assay ≥98%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00051885
Beilstein 4281936
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 333° to 335°C
Health Hazard 1 Warning
Synonym 1,3-Bis(triphenylphosphonio)propane dibromide
Chemical Name or Material Trimethylenebis(triphenylphosphonium bromide)
CAS 7333-67-7
Molecular Formula C39H36Br2P2
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.