Please login to your online account to display your discounted pricing

Alfa Aesar™ Trimethylene borate, 98%

$204.73 - $204.73

Chemical Identifiers

Chemical Name or Material Trimethylene borate
CAS 20905-35-5
Molecular Formula C9H18B2O6
Formula Weight 243.86
MDL Number MFCD03701690
Beilstein 1913020
Synonym Boric acid cyclic ester with 1,3-propanediol; Boric acid 1,3-propanediol cyclic ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2193314

  SDS
alfa aesar™
B2193314
25g Each for $204.73
Description & Specifications

Specifications

Formula Weight 243.86
Assay 98%
Molecular Formula C9H18B2O6
MDL Number MFCD03701690
CAS 20905-35-5
Beilstein 1913020
Synonym Boric acid cyclic ester with 1,3-propanediol; Boric acid 1,3-propanediol cyclic ester
Flash Point >110°C (230°F)
Boiling Point 145° to 148°C (0.2mmHg)
Quantity 25g
Chemical Name or Material Trimethylene borate