missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trimethylamine, pure, 4.2M (33 wt.%) solution in ethanol, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 270442500

 View more versions of this product

Catalog No. AC270442500



Description

Description

Specifications

Specifications

Trimethylamine
−20°C
1.3630 to 1.3660
50.0°C
250mL
C3H9N
MFCD00008327
15,988
Solubility in water: soluble. Other solubilities: soluble in alcohol,ether,benzene,toluene,,xylene,ethylbenzene and chloroform
CN(C)C
59.11
CHEBI:18139
Liquid
Pure
0.7500g/mL
Glass bottle
0.75
Colorless
75-50-3,64-17-5
(CH3)3N
04,43
ch3 3n, dimethylmethaneamine, fema number 3241, methanamine, n,n-dimethyl, n-trimethylamine, trimethyl amine, trimethyl-amine, trimethylamin, trimethylamine, trimethylamine solution
GETQZCLCWQTVFV-UHFFFAOYSA-N
N,N-dimethylmethanamine
1146
59.11
32 to 34%

Chemical Identifiers

75-50-3
59.11
GETQZCLCWQTVFV-UHFFFAOYSA-N
1146
N,N-dimethylmethanamine
C3H9N
MFCD00008327
ch3 3n, dimethylmethaneamine, fema number 3241, methanamine, n,n-dimethyl, n-trimethylamine, trimethyl amine, trimethyl-amine, trimethylamin, trimethylamine, trimethylamine solution
CHEBI:18139
CN(C)C
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Highly flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 200-875-0

RTECSNumber : PA0350000

TSCA : TSCA

SDS Documents