Learn More
Trimethylamine, 33% w/w in ethanol denatured with 2% cyclohexane, Thermo Scientific Chemicals
$501.19 - $501.19
Chemical Identifiers
CAS | 75-50-3 |
---|---|
Molecular Formula | C3H9N |
Molecular Weight (g/mol) | 59.11 |
MDL Number | MFCD00008327 |
InChI Key | GETQZCLCWQTVFV-UHFFFAOYSA-N |
Synonym | trimethylamine, methanamine, n,n-dimethyl, dimethylmethaneamine, n-trimethylamine, trimethylamine solution, ch3 3n, trimethyl amine, trimethylamin, trimethyl-amine, fema number 3241 |
PubChem CID | 1146 |
ChEBI | CHEBI:18139 |
IUPAC Name | N,N-dimethylmethanamine |
SMILES | CN(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH27324AU
|
Thermo Scientific Chemicals
H27324AU |
1000 mL |
Each for $501.19
|
|
|||||
Description
Mainly as intermediate used in the production of industrial chemicals, initial product for chemical synthesis, dyes, textile industry, active ingredient. Trimethylamine is used in the synthesis of choline, tetramethylammonium hydroxide, plant growth regulators.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMainly as intermediate used in the production of industrial chemicals, initial product for chemical synthesis, dyes, textile industry, active ingredient. Trimethylamine is used in the synthesis of choline, tetramethylammonium hydroxide, plant growth regulators.
Solubility
Insoluble in water.
Notes
Air Sensitive & Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, water, moisture, acids, brass, zinc, magnesium, aluminum, mercury, mercury oxides, acid chlorides, acid anhydrides, strong reducing agents.
Chemical Identifiers
75-50-3 | |
59.11 | |
GETQZCLCWQTVFV-UHFFFAOYSA-N | |
1146 | |
N,N-dimethylmethanamine |
C3H9N | |
MFCD00008327 | |
trimethylamine, methanamine, n,n-dimethyl, dimethylmethaneamine, n-trimethylamine, trimethylamine solution, ch3 3n, trimethyl amine, trimethylamin, trimethyl-amine, fema number 3241 | |
CHEBI:18139 | |
CN(C)C |
Specifications
75-50-3 | |
−20°C (−4°F) | |
1.3635 | |
MFCD00008327 | |
UN2924 | |
Air sensitive; Hygroscopic | |
trimethylamine, methanamine, n,n-dimethyl, dimethylmethaneamine, n-trimethylamine, trimethylamine solution, ch3 3n, trimethyl amine, trimethylamin, trimethyl-amine, fema number 3241 | |
GETQZCLCWQTVFV-UHFFFAOYSA-N | |
N,N-dimethylmethanamine | |
1146 | |
59.11 | |
Trimethylamine |
0.75 | |
C3H9N | |
(CH3)3N | |
1000 mL | |
956566 | |
14,9710 | |
Insoluble in water. | |
CN(C)C | |
59.11 | |
CHEBI:18139 | |
33% w/w in ethanol |
Safety and Handling
GHS H Statement
H225-H314-H302-H335
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H302+H332-H304-H314-H335-H336
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber : 200-875-0
TSCA : Yes
Recommended Storage : Ambient temperatures
For Manufacturing and Laboratory Use. Not for beverage use