Please login to your online account to display your discounted pricing

Trimethylamine, 24% Aqueous Solution, ACROS Organics™

$34.34 - $39.30

Chemical Identifiers

Chemical Name or Material Trimethylamine
CAS 75-50-3
CAS 7732-18-5
Purity Grade Extra Pure
Assay 23 to 25%
Molecular Formula C3H9N
Linear Formula (CH3)3N
Formula Weight 59.11
MDL Number MFCD00008327
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC421870250

  SDS
thermo fisher scientific
421870250
Glass bottle 25g Each for $34.34

AC421875000

  SDS
thermo fisher scientific
421875000
Glass bottle 500g Each for $39.30
Description & Specifications

Specifications

Purity Grade Extra Pure
Formula Weight 59.11
Physical Form Liquid
Quantity 25g
Color Clear colorless to slightly yellow
Assay 23 to 25%
RTECS Number PA0350000
TSCA TSCA
EINECS Number 200-875-0
Density 0.92g/mL
Beilstein 04, 43
Merck Index 15, 9880
CAS 7732-18-5
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Linear Formula (CH3)3N
Chemical Name or Material Trimethylamine
CAS 75-50-3
Molecular Formula C3H9N
MDL Number MFCD00008327
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Highly flammable liquid and vapor.