Learn More
Trimethylacetonitrile, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1201122
Description
Trimethylacetonitrile used as a solvent and as a labile ligand in coordination chemistry. It is also used as an intermediate in organic synthesis. It is used in the presence of titanium(II) chloride and zinc to undergoes reductive coupling reaction of aromatic and aliphatic ketones including acyclic aliphatic ketones to give the corresponding pinacols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTrimethylacetonitrile used as a solvent and as a labile ligand in coordination chemistry. It is also used as an intermediate in organic synthesis. It is used in the presence of titanium(II) chloride and zinc to undergoes reductive coupling reaction of aromatic and aliphatic ketones including acyclic aliphatic ketones to give the corresponding pinacols.
Solubility
Miscible with ethanol, acetone and toluene. Slightly miscible with water.
Notes
Store in a cool place. Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents.
Specifications
Trimethylacetonitrile | |
16°C to 18°C | |
105°C to 106°C | |
Odorless | |
MFCD00001847 | |
UN3273 | |
Miscible with ethanol,acetone and toluene. Slightly miscible with water. | |
CC(C)(C)C#N | |
83.134 | |
83.13 |
630-18-2 | |
0.757 | |
4°C (39°F) | |
C5H9N | |
100 g | |
pivalonitrile, trimethylacetonitrile, 2,2-dimethylpropionitrile, tert-butyl cyanide, propanenitrile, 2,2-dimethyl, tert-butylnitrile, 2-cyano-2-methylpropane, trimethyl acetonitrile, tert-cyanobutane, t-bucn | |
JAMNHZBIQDNHMM-UHFFFAOYSA-N | |
2,2-dimethylpropanenitrile | |
12416 | |
≥98% |
Chemical Identifiers
630-18-2 | |
83.134 | |
JAMNHZBIQDNHMM-UHFFFAOYSA-N | |
12416 | |
CC(C)(C)C#N |
C5H9N | |
MFCD00001847 | |
pivalonitrile, trimethylacetonitrile, 2,2-dimethylpropionitrile, tert-butyl cyanide, propanenitrile, 2,2-dimethyl, tert-butylnitrile, 2-cyano-2-methylpropane, trimethyl acetonitrile, tert-cyanobutane, t-bucn | |
2,2-dimethylpropanenitrile |
Safety and Handling
GHS H Statement
H225-H302-H312-H332-H315-H319-H335
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P311-P312-P330-P363-P370+P378q-P501c
H225-H301+H311+H331
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: NITRILES, FLAMMABLE, TOXIC, N.O.S.
EINECSNumber : 211-133-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only