Learn More
Trimethyl thiophosphate, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03039503
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Trimethyl thiophosphate | |
White | |
C3H9O3PS | |
1 g | |
trimethyl thiophosphate, trimethylthiophosphate, o,o,o-trimethyl phosphorothioate, o,o,o-trimethyl thiophosphate, trimethylthiofosfat, o,o,o-trimethylthiofosfat, trimethylthiofosfat czech, ccris 5569, phosphorothioic acid, o,o,o-trimethyl ester, o,o,o-trimethylthiofosfat czech | |
COP(=S)(OC)OC | |
156.136 | |
156.15 |
152-18-1 | |
Sulphurous | |
MFCD00014888 | |
UN3278 | |
XWSLYQXUTWUIKM-UHFFFAOYSA-N | |
trimethoxy(sulfanylidene)-$l^{5}-phosphane | |
9038 | |
Liquid |
Chemical Identifiers
152-18-1 | |
156.136 | |
XWSLYQXUTWUIKM-UHFFFAOYSA-N | |
9038 | |
COP(=S)(OC)OC |
C3H9O3PS | |
MFCD00014888 | |
trimethyl thiophosphate, trimethylthiophosphate, o,o,o-trimethyl phosphorothioate, o,o,o-trimethyl thiophosphate, trimethylthiofosfat, o,o,o-trimethylthiofosfat, trimethylthiofosfat czech, ccris 5569, phosphorothioic acid, o,o,o-trimethyl ester, o,o,o-trimethylthiofosfat czech | |
trimethoxy(sulfanylidene)-$l^{5}-phosphane |
Safety and Handling
GHS H Statement
H330-H302-H315-H319-H335
Fatal if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P270-P271-P280-P284-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H330-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, LIQUID, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only