missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Trimethyl phosphite, 99%

Manufacturer:  Alfa Aesar™ 4398830

 View more versions of this product

Catalog No. AA4398830



Specifications

Specifications

Trimethyl phosphite
27°C (80°F)
1.4080
111°C to 112°C
-78°C
121-45-9
(CH3O)3P
UN2329
CYTQBVOFDCPGCX-UHFFFAOYSA-N
trimethyl phosphite
8472
Liquid
1.052g/mL
Unpleasant
Air & Moisture Sensitive
Colorless
250g
C3H9O3P
MFCD00008350
trimethoxyphosphine, phosphorous acid, trimethyl ester, trimethylphosphite, methyl phosphite, trimethylfosfit, trimethoxyfosfin, trimethylfosfit czech, trimethoxyfosfin czech, fosforyn trojmetylowy czech, unii-26q0321zdg
COP(OC)OC
124.076
124.08
99%

Chemical Identifiers

121-45-9
124.076
CYTQBVOFDCPGCX-UHFFFAOYSA-N
8472
COP(OC)OC
C3H9O3P
MFCD00008350
trimethoxyphosphine, phosphorous acid, trimethyl ester, trimethylphosphite, methyl phosphite, trimethylfosfit, trimethoxyfosfin, trimethylfosfit czech, trimethoxyfosfin czech, fosforyn trojmetylowy czech, unii-26q0321zdg
trimethyl phosphite
Safety and Handling

Safety and Handling

GHS H Statement
H226-H361-H302-H315-H319-H335
Flammable liquid and vapor.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: TRIMETHYL PHOSPHITE

TSCA : TSCA

Documents