missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trimethyl phosphite, 97%, Alfa Aesar™

Manufacturer:  Alfa Aesar 3013222

 View more versions of this product

Catalog No. AA3013222



Specifications

Specifications

Trimethyl phosphite
100g
97%
(CH3O)3P
UN2329
CYTQBVOFDCPGCX-UHFFFAOYSA-N
trimethyl phosphite
8472
Liquid
1.052g/mL
121-45-9
C3H9O3P
MFCD00008350
trimethoxyphosphine, phosphorous acid, trimethyl ester, trimethylphosphite, methyl phosphite, trimethylfosfit, trimethoxyfosfin, trimethylfosfit czech, trimethoxyfosfin czech, fosforyn trojmetylowy czech, unii-26q0321zdg
COP(OC)OC
124.076
124.08
97%

Chemical Identifiers

121-45-9
124.076
CYTQBVOFDCPGCX-UHFFFAOYSA-N
8472
COP(OC)OC
C3H9O3P
MFCD00008350
trimethoxyphosphine, phosphorous acid, trimethyl ester, trimethylphosphite, methyl phosphite, trimethylfosfit, trimethoxyfosfin, trimethylfosfit czech, trimethoxyfosfin czech, fosforyn trojmetylowy czech, unii-26q0321zdg
trimethyl phosphite
Safety and Handling

Safety and Handling

GHS H Statement
H226-H361-H302-H315-H319-H335

GHS P Statement
P210-P201-P261-P280-P281-P303+P361+P353-P305+P351+P338-P304+P340-P405-P501a

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: TRIMETHYL PHOSPHITE

RTECSNumber : TH1400000

TSCA : TSCA

SDS Documents