Learn More
Trimethyl phosphite, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03013236
Description
It is an important raw material and intermediate used in Organic synthesis..
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Trimethyl phosphite | |
-78°C | |
1.052 g/mL | |
27°C (80°F) | |
Unpleasant | |
1.408 | |
MFCD00008350 | |
UN2329 | |
trimethoxyphosphine, phosphorous acid, trimethyl ester, trimethylphosphite, methyl phosphite, trimethylfosfit, trimethoxyfosfin, trimethylfosfit czech, trimethoxyfosfin czech, fosforyn trojmetylowy czech, unii-26q0321zdg | |
CYTQBVOFDCPGCX-UHFFFAOYSA-N | |
trimethyl phosphite | |
8472 | |
97% |
121-45-9 | |
Colorless | |
111°C to 112°C | |
97% | |
C3H9O3P | |
(CH3O)3P | |
500 g | |
Air and moisture sensitive | |
Hydrolyzes with water. | |
COP(OC)OC | |
124.076 | |
124.08 | |
Liquid |
Chemical Identifiers
121-45-9 | |
124.076 | |
CYTQBVOFDCPGCX-UHFFFAOYSA-N | |
8472 | |
COP(OC)OC |
C3H9O3P | |
MFCD00008350 | |
trimethoxyphosphine, phosphorous acid, trimethyl ester, trimethylphosphite, methyl phosphite, trimethylfosfit, trimethoxyfosfin, trimethylfosfit czech, trimethoxyfosfin czech, fosforyn trojmetylowy czech, unii-26q0321zdg | |
trimethyl phosphite |
Safety and Handling
GHS H Statement
H226-H361-H302-H315-H319-H335
Flammable liquid and vapor.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P363-P370+P378q-P501c
H226-H302+H312+H332
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: TRIMETHYL PHOSPHITE
EINECSNumber : 204-471-5
RTECSNumber : TH1400000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only