Please login to your online account to display your discounted pricing

Alfa Aesar™ Trimethyl orthovalerate, 98%

Chemical Identifiers

Chemical Name or Material Trimethyl orthovalerate
CAS 13820-09-2
Molecular Formula C8H18O3
Linear Formula CH3(CH2)3C(OCH3)3
Formula Weight 162.23
MDL Number MFCD00008481
UN Number UN1993
Beilstein 1739200
Merck Index 14,9717
Synonym Orthovaleric acid trimethyl ester; 1,1,1-Trimethoxypentane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2037706

  SDS
alfa aesar™
L2037706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL2037714

  SDS
alfa aesar™
L2037714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 162.23
MDL Number MFCD00008481
CAS 13820-09-2
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
TSCA TSCA
Assay 98%
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 164° to 166°C
Quantity 5g
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Linear Formula CH3(CH2)3C(OCH3)3
Synonym Orthovaleric acid trimethyl ester; 1,1,1-Trimethoxypentane
Beilstein 1739200
Merck Index 14,9717
UN Number UN1993
Flash Point 41°C (105°F)
Molecular Formula C8H18O3
Chemical Name or Material Trimethyl orthovalerate
Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.