missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trimethyl Orthopropionate, TCI America™

Manufacturer:  TCI America T228825G

 View more versions of this product

Catalog No. T228825G



Specifications

Specifications

Trimethyl Orthopropionate
Colorless
24823-81-2
MFCD00043305
trimethyl orthopropionate, propane, 1,1,1-trimethoxy, trimethylorthopropionate, trimethoxypropane, acmc-209gew, trimethyl orthopropanoate, propane,1,1,1-trimethoxy, ksc489s6b, zgmnaiodrdomek-uhfffaoysa, orthopropionic acid trimethyl ester
CCC(OC)(OC)OC
134.175
134.18
≥97.0% (GC)
128°C
25g
C6H14O3
3272
ZGMNAIODRDOMEK-UHFFFAOYSA-N
1,1,1-trimethoxypropane
141145
Clear Liquid at 20°C

Chemical Identifiers

24823-81-2
134.175
ZGMNAIODRDOMEK-UHFFFAOYSA-N
141145
CCC(OC)(OC)OC
C6H14O3
MFCD00043305
trimethyl orthopropionate, propane, 1,1,1-trimethoxy, trimethylorthopropionate, trimethoxypropane, acmc-209gew, trimethyl orthopropanoate, propane,1,1,1-trimethoxy, ksc489s6b, zgmnaiodrdomek-uhfffaoysa, orthopropionic acid trimethyl ester
1,1,1-trimethoxypropane
Safety and Handling

Safety and Handling

TSCA : No

Documents