Please login to your online account to display your discounted pricing

Trimethyl orthopropionate 97%, ACROS Organics™


Quantity 100mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC384621000

 View more versions of this product

Catalog No. AC384621000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 134.18
Quantity 100mL
Color Clear colorless
Density 0.944g/mL
Chemical Name or Material Trimethyl orthopropionate
Molecular Formula C6H14O3
MDL Number MFCD00043305
CAS 24823-81-2
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.3970 to 1.3990 (20°C, 589nm)
Assay Percent Range 97%
Linear Formula CH3CH2C(OCH3)3
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.

Specifications

Chemical Name or Material Trimethyl orthopropionate
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H14O3
Linear Formula CH3CH2C(OCH3)3
Formula Weight 134.18
MDL Number MFCD00043305
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.

SDS & Safety Documents