missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trimethyl orthopropionate 97%, ACROS Organics™

$214.74 - $214.74

Chemical Identifiers

Assay Percent Range 97%
Molecular Formula C6H14O3
Linear Formula CH3CH2C(OCH3)3
Formula Weight 134.18
MDL Number MFCD00043305
Chemical Name or Material Trimethyl orthopropionate
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC384621000 SDS Thermo Fisher Scientific
AC384621000
Glass bottle 100mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC384620250 SDS Thermo Fisher Scientific
AC384620250
Glass bottle 25mL Each for $214.74
Specifications Description & Specifications

Chemical Identifiers

97%
CH3CH2C(OCH3)3
MFCD00043305
≥96.0%
C6H14O3
134.18
Trimethyl orthopropionate

Specifications

0.944g/mL
100mL
Authentic
1.3970 to 1.3990 (20°C, 589nm)
97%
CH3CH2C(OCH3)3
MFCD00043305
≥96.0%
Clear colorless
24823-81-2
Glass bottle
Liquid
C6H14O3
134.18
Trimethyl orthopropionate

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger