Please login to your online account to display your discounted pricing

Trimethyl orthobutyrate 97%, ACROS Organics™


Quantity 50mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC384630500

 View more versions of this product

Catalog No. AC384630500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 148.2
Quantity 50mL
Color Clear colorless to slightly yellow
Density 0.932g/mL
Chemical Name or Material Trimethyl orthobutyrate
Assay Percent Range 97%
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4020 to 1.4040 (20°C, 589nm)
Molecular Formula C7H16O3
Linear Formula CH3(CH2)2C(OCH3)3
MDL Number MFCD00008480
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
CAS 43083-12-1

Specifications

Chemical Name or Material Trimethyl orthobutyrate
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H16O3
Linear Formula CH3(CH2)2C(OCH3)3
Formula Weight 148.2
MDL Number MFCD00008480
Accessories

GHS Signal Word: Warning

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

SDS & Safety Documents