Please login to your online account to display your discounted pricing

Trimethyl orthobutyrate 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Trimethyl orthobutyrate
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H16O3
Linear Formula CH3(CH2)2C(OCH3)3
Formula Weight 148.2
MDL Number MFCD00008480
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC384630500

  SDS
thermo fisher scientific
AC384630500
Glass bottle 50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 148.2
Quantity 50mL
Color Clear colorless to slightly yellow
Density 0.932g/mL
Infrared Spectrum Authentic
Chemical Name or Material Trimethyl orthobutyrate
Health Hazard 1 GHS Signal Word: Warning
Assay Percent Range 97%
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00008480
Refractive Index 1.4020 to 1.4040 (20°C, 589nm)
Linear Formula CH3(CH2)2C(OCH3)3
Packaging Glass bottle
Molecular Formula C7H16O3
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
CAS 43083-12-1