Please login to your online account to display your discounted pricing

Trimethyl orthobenzoate, 97%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  172920100

 View more versions of this product

Catalog No. AC172920100

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 182.22
Quantity 10g
Color Clear colorless
EINECS Number 211-897-5
TSCA TSCA
Density 1.06g/mL
Fieser 13,327; 04,548; 13,195; 01,1256; 02,454; 04,565; 06,657; 07,414; 08,524; 09,512; 10,455; 13,301
Chemical Name or Material Trimethyl orthobenzoate
Packaging Glass bottle
Refractive Index 1.4880 to 1.4900 (20°C, 589nm)
Name Note 97%
Assay Percent Range 97%
Molecular Formula C10H14O3
Linear Formula C6H5C(OCH3)3
MDL Number MFCD00008474
CAS 707-07-3
Health Hazard 1 MOISTURE SENSITIVE
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Trimethyl orthobenzoate
Name Note 97%
CAS 707-07-3
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H14O3
Linear Formula C6H5C(OCH3)3
Formula Weight 182.22
MDL Number MFCD00008474
Accessories

MOISTURE SENSITIVE

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents