Please login to your online account to display your discounted pricing

(Trimethyl)methylcyclopentadienylplatinum(IV), 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (Trimethyl)methylcyclopentadienylplatinum(IV)
CAS 94442-22-5
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C9H16Pt
Formula Weight 319.32
MDL Number MFCD00079665
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC363631000

  SDS
fisher scientific acros
363631000
Glass bottle 100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 319.32
Physical Form Crystalline Powder or Crystals
Quantity 100mg
Color White to off-white
TSCA TSCA
Melting Point 29° to 32°C
CAS Max % 100.0
CAS Min % 98.5
Packaging Glass bottle
Infrared Spectrum Authentic
NMR 98.5% min.
Health Hazard 1 AIR SENSITIVE
Chemical Name or Material (Trimethyl)methylcyclopentadienylplatinum(IV)
Molecular Formula C9H16Pt
CAS 94442-22-5
Assay Percent Range 99%
MDL Number MFCD00079665
Safety and Handling

AIR SENSITIVE