Learn More
Trimethyl borate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 140121000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Trimethyl borate | |
121-43-7 | |
Colorless | |
68.0°C to 69.0°C | |
Authentic | |
Glass bottle | |
1.3562 to 1.3582 | |
MFCD00008346 | |
01, 287 | |
0.915 | |
trimethoxyborane, methyl borate, trimethoxyboron, boric acid, trimethyl ester, trimethylborate, boric acid trimethyl ester, trimethoxyborine, borester o, boric acid h3bo3 , trimethyl ester, b och3 3 | |
WRECIMRULFAWHA-UHFFFAOYSA-N | |
trimethyl borate | |
8470 | |
CHEBI:38913 | |
99% |
99% | |
-34.0°C | |
0.9150g/mL | |
−8°C | |
99% | |
C3H9O3B | |
(CH3O)3B | |
100 g | |
01,673; 13,218 | |
15, 9882 | |
Solubility in water: reacts. Other solubilities: miscible with thf,diethyl ether,hexane,methanol,,isopropylamine,and other organic liquids | |
B(OC)(OC)OC | |
103.91 | |
0.36 mPa.s (25°C) | |
103.91 | |
Liquid |
Chemical Identifiers
121-43-7 | |
WRECIMRULFAWHA-UHFFFAOYSA-N | |
8470 | |
trimethyl borate |
MFCD00008346 | |
trimethoxyborane, methyl borate, trimethoxyboron, boric acid, trimethyl ester, trimethylborate, boric acid trimethyl ester, trimethoxyborine, borester o, boric acid h3bo3 , trimethyl ester, b och3 3 | |
CHEBI:38913 | |
B(OC)(OC)OC |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Highly flammable liquid and vapor.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 204-468-9
RTECSNumber : ED5600000
TSCA : TSCA
RUO – Research Use Only