missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trimethyl borate, 99.9995+% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 2297530

 View more versions of this product

Catalog No. AA2297530



Specifications

Specifications

Trimethyl borate
−8° (17°F)
1.3610
67°C to 69°C
-34°C
121-43-7
(metals basis)
(CH3O)3B
UN2416
14,9712
WRECIMRULFAWHA-UHFFFAOYSA-N
trimethyl borate
8470
103.92
≥99.9995%
0.915g/mL
Odorless
Moisture Sensitive
Colorless
250g
>99.9995% (metals basis)
C3H9BO3
MFCD00008346
1697939
trimethoxyborane, methyl borate, trimethoxyboron, boric acid, trimethyl ester, trimethylborate, boric acid trimethyl ester, trimethoxyborine, borester o, boric acid h3bo3 , trimethyl ester, b och3 3
B(OC)(OC)OC
103.912
CHEBI:38913
Liquid

Chemical Identifiers

121-43-7
103.912
WRECIMRULFAWHA-UHFFFAOYSA-N
8470
trimethyl borate
C3H9BO3
MFCD00008346
trimethoxyborane, methyl borate, trimethoxyboron, boric acid, trimethyl ester, trimethylborate, boric acid trimethyl ester, trimethoxyborine, borester o, boric acid h3bo3 , trimethyl ester, b och3 3
CHEBI:38913
B(OC)(OC)OC
Safety and Handling

Safety and Handling

GHS H Statement
H225-H312
Highly flammable liquid and vapor.
Harmful in contact with skin.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TRIMETHYL BORATE

RTECSNumber : ED5600000

TSCA : TSCA

SDS Documents