Please login to your online account to display your discounted pricing

Alfa Aesar™ Trimethyl borate, 70% azeotrope with methanol

Chemical Identifiers

Chemical Name or Material Trimethyl borate
CAS 121-43-7
Molecular Formula (CH3O)3B
Formula Weight 103.92
MDL Number MFCD00008346
UN Number UN1993
Merck Index 14,9712
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA44721AK

 
alfa aesar™
44721AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA44721K2

 
alfa aesar™
44721K2
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA44721K7

 
alfa aesar™
44721K7
4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 103.92
Chemical Name or Material Trimethyl borate
TSCA TSCA
Physical Form Liquid
Molecular Formula (CH3O)3B
MDL Number MFCD00008346
CAS 121-43-7
UN Number UN1993
Merck Index 14,9712
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H330-H370-H226-H312
Fatal if swallowed.
Fatal if inhaled.
Causes damage to organs.
Flammable liquid and vapor.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P304+P340-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Concentration 70% azeotrope with methanol
Density 0.885g/mL
Flash Point −9°C (16°F)
Odor Characteristic
Boiling Point 52° to 54°C
Quantity 250mL
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P304+P340-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H330-H370-H226-H312
Fatal if swallowed.
Fatal if inhaled.
Causes damage to organs.
Flammable liquid and vapor.
Harmful in contact with skin.