missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Trimethyl[3-(triethoxysilyl)propyl]ammonium Chloride 98.0+%, TCI America™
Supplier: TCI America T224625G
Specifications
Trimethyl[3-(triethoxysilyl)propyl]ammonium Chloride | |
White-Yellow | |
MFCD00043316 | |
MVYLHRSUMWTUJE-UHFFFAOYNA-M | |
trimethyl[1-(triethoxysilyl)propyl]azanium chloride | |
23002981 | |
≥98.0% (T) |
84901-27-9 | |
C12H30ClNO3Si | |
25 g | |
[Cl-].CCO[Si](OCC)(OCC)C(CC)[N+](C)(C)C | |
299.91 | |
299.91 | |
Crystalline Powder |
Chemical Identifiers
84901-27-9 | |
299.91 | |
MVYLHRSUMWTUJE-UHFFFAOYNA-M | |
trimethyl[1-(triethoxysilyl)propyl]azanium chloride |
C12H30ClNO3Si | |
MFCD00043316 | |
23002981 | |
[Cl-].CCO[Si](OCC)(OCC)C(CC)[N+](C)(C)C |
Safety and Handling
EINECSNumber : (2)-2094&(2)-2095
TSCA : No