missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trimethyl[2,3-(dioleyloxy)propyl]ammonium Chloride, TCI America™

Manufacturer:  TCI America T1307100MG

 View more versions of this product

Catalog No. T1307100MG



Specifications

Specifications

Trimethyl[2,3-(dioleyloxy)propyl]ammonium Chloride
100mg
C42H84ClNO2
LDGWQMRUWMSZIU-AFLLVNNISA-M
2,3-bis[(E)-octadec-9-enoxy]propyl-trimethylazanium;chloride
16730297
Crystal-Lump at 20°C
Undesignated
104162-48-3
N-[1-(2,3-Dioleyloxy)propyl]-N,N,N-trimethylammonium Chloride, DOTMA
CCCCCCCCC=CCCCCCCCCOCC(C[N+](C)(C)C)OCCCCCCCCC=CCCCCCCCC.[Cl-]
670.589
670.59
≥90.0% (T)

Chemical Identifiers

104162-48-3
670.589
N-[1-(2,3-Dioleyloxy)propyl]-N,N,N-trimethylammonium Chloride, DOTMA
2,3-bis[(E)-octadec-9-enoxy]propyl-trimethylazanium;chloride
C42H84ClNO2
LDGWQMRUWMSZIU-AFLLVNNISA-M
16730297
CCCCCCCCC=CCCCCCCCCOCC(C[N+](C)(C)C)OCCCCCCCCC=CCCCCCCCC.[Cl-]
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents