missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Trimethyl 1,2,4-benzenetricarboxylate, 98%

$18.32 - $153.16

Chemical Identifiers

CAS 2459-10-1
Molecular Formula C12H12O6
Formula Weight 252.22
MDL Number MFCD00043629
Synonym 1,2,4-Benzenetricarboxylic acid trimethyl ester; Trimethyl trimellitate
Chemical Name or Material Trimethyl 1,2,4-benzenetricarboxylate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2445514 SDS Alfa Aesar™
B2445514
25g Each for $18.32
AAB2445522 SDS Alfa Aesar™
B2445522
100g Each for $44.01
AAB2445536 SDS Alfa Aesar™
B2445536
500g Each for $153.16
Specifications Description & Specifications

Chemical Identifiers

2459-10-1
252.22
1,2,4-Benzenetricarboxylic acid trimethyl ester; Trimethyl trimellitate
C12H12O6
MFCD00043629
Trimethyl 1,2,4-benzenetricarboxylate

Specifications

182°C (360°F)
38° to 40°C
2459-10-1
252.22
1,2,4-Benzenetricarboxylic acid trimethyl ester; Trimethyl trimellitate
98%
193° to 195°C (12mmHg)
25g
C12H12O6
MFCD00043629
Trimethyl 1,2,4-benzenetricarboxylate

Safety and Handling

TSCA : TSCA