Please login to your online account to display your discounted pricing

Trimethoxysilane 95%, ACROS Organics™

$183.07 - $525.27

Chemical Identifiers

Chemical Name or Material Trimethoxysilane
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C3H10O3Si
Linear Formula HSi(OCH3)3
Formula Weight 122.2
MDL Number MFCD00008340
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC392170250

  SDS
thermo fisher scientific
AC392170250
Glass bottle 25g Each for $183.07

AC392170050

  SDS
thermo fisher scientific
AC392170050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC392171000

  SDS
thermo fisher scientific
AC392171000
Glass bottle 100g Each for $525.27
Description & Specifications

Specifications

Assay ≥94%
Physical Form Liquid
Formula Weight 122.2
Quantity 25g
Color Clear colorless
Density 0.8600g/mL
Chemical Name or Material Trimethoxysilane
Packaging Glass bottle
Refractive Index 1.3570 to 1.3600 (20°C, 589nm)
Assay Percent Range 95%
Molecular Formula C3H10O3Si
Linear Formula HSi(OCH3)3
MDL Number MFCD00008340
CAS 2487-90-3
Health Hazard 3 GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Fatal if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Highly flammable liquid and vapor.

Reduction reagent

Safety and Handling

Danger

GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Fatal if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Highly flammable liquid and vapor.