missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Trimethoxyboroxine

Manufacturer:  Alfa Aesar™ 8915314

 View more versions of this product

Catalog No. AA8915314



Specifications

Specifications

Trimethoxyboroxine
14°C (57°F)
1.3996
130°C
25g
C3H9B3O6
UN2924
trimethoxyboroxine, trimethoxyboroxin, boroxin, trimethoxy, trimethoxycyclotriboroxane, 2,4,6-trimethoxyboroxin, triboron trimethyl hexaoxide, boroxin, 2,4,6-trimethoxy, boric acid h3b3o6 , trimethyl ester, trimethoxy-boroxi, trimethoxyboroxole
B1(OB(OB(O1)OC)OC)OC
173.529
173.53
1.216g/mL
Strong
Air & Moisture Sensitive
10°C to 11°C
102-24-9
MFCD00005946
1709158
ZFMOJHVRFMOIGF-UHFFFAOYSA-N
2,4,6-trimethoxy-1,3,5,2,4,6-trioxatriborinane
66880
Liquid

Chemical Identifiers

102-24-9
173.529
ZFMOJHVRFMOIGF-UHFFFAOYSA-N
66880
B1(OB(OB(O1)OC)OC)OC
C3H9B3O6
MFCD00005946
trimethoxyboroxine, trimethoxyboroxin, boroxin, trimethoxy, trimethoxycyclotriboroxane, 2,4,6-trimethoxyboroxin, triboron trimethyl hexaoxide, boroxin, 2,4,6-trimethoxy, boric acid h3b3o6 , trimethyl ester, trimethoxy-boroxi, trimethoxyboroxole
2,4,6-trimethoxy-1,3,5,2,4,6-trioxatriborinane
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

RTECSNumber : ED8720000

TSCA : TSCA

SDS Documents