Please login to your online account to display your discounted pricing

Alfa Aesar™ Trimethoxy(pentamethylcyclopentadienyl)titanium(IV), 97+%

$147.51 - $452.03

Chemical Identifiers

Chemical Name or Material Trimethoxy(pentamethylcyclopentadienyl)titanium(IV)
CAS 123927-75-3
Molecular Formula C13H24O3Ti
Formula Weight 276.21
MDL Number MFCD00269850
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2788503

  SDS
alfa aesar™
H2788503
1g Each for $147.51

AAH2788506

  SDS
alfa aesar™
H2788506
5g Each for $452.03
Description & Specifications

Specifications

Formula Weight 276.21
Health Hazard 1 Warning
Chemical Name or Material Trimethoxy(pentamethylcyclopentadienyl)titanium(IV)
CAS 123927-75-3
MDL Number MFCD00269850
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Quantity 1g
Boiling Point 285°C
Flash Point 61°C (142°F)
Molecular Formula C13H24O3Ti
Assay ≥97%
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.