missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trimethoxy(1H,1H,2H,2H-nonafluorohexyl)silane, TCI America™

Manufacturer:  TCI America T29185G

 View more versions of this product

Catalog No. T29185G



Specifications

Specifications

Trimethoxy(1H,1H,2H,2H-nonafluorohexyl)silane
Colorless
85877-79-8
MFCD08275518
IJROHELDTBDTPH-UHFFFAOYSA-N
trimethoxy(3,3,4,4,5,5,6,6,6-nonafluorohexyl)silane
15651659
Clear Liquid at 20°C
43°C
5g
C9H13F9O3Si
Trimethoxy(1H,1H,2H,2H-perfluorohexyl)silane, 1H,1H,2H,2H-Nonafluorohexyltrimethoxysilane
CO[Si](CCC(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(OC)OC
368.271
368.27
≥97.0% (GC)

Chemical Identifiers

85877-79-8
368.271
IJROHELDTBDTPH-UHFFFAOYSA-N
15651659
CO[Si](CCC(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(OC)OC
C9H13F9O3Si
MFCD08275518
Trimethoxy(1H,1H,2H,2H-perfluorohexyl)silane, 1H,1H,2H,2H-Nonafluorohexyltrimethoxysilane
trimethoxy(3,3,4,4,5,5,6,6,6-nonafluorohexyl)silane
Safety and Handling

Safety and Handling

TSCA : No

Documents