Please login to your online account to display your discounted pricing

Alfa Aesar™ Trimethoprim lactate, 20mg/mL in distilled water, sterile-filtered

Chemical Identifiers

Chemical Name or Material Trimethoprim lactate
CAS 23256-42-0
Name Note 20mg/mL in distilled water, sterile-filtered
MDL Number MFCD00171722
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ673958EQ

 
alfa aesar™
J673958EQ
5 x 1mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
MDL Number MFCD00171722
CAS 23256-42-0
Name Note 20mg/mL in distilled water, sterile-filtered
Storage Note 1 -30° to -10°C
Chemical Name or Material Trimethoprim lactate
Quantity 5 x 1mL
Safety and Handling

-30° to -10°C