Please login to your online account to display your discounted pricing

Alfa Aesar™ Trimethoprim, 50mg/mL in DMSO, sterile-filtered

$232.43 - $232.43

Chemical Identifiers

Chemical Name or Material Trimethoprim
CAS 738-70-5
Name Note 50mg/mL in DMSO, sterile-filtered
MDL Number MFCD00036761
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ671758EQ

 
alfa aesar™
J671758EQ
5 x 1mL Pack for $232.43
Description & Specifications

Specifications

Chemical Name or Material Trimethoprim
CAS 738-70-5
Name Note 50mg/mL in DMSO, sterile-filtered
Physical Form Liquid
MDL Number MFCD00036761
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 -30° to -10°C
RTECS Number UV8225000
Quantity 5 x 1mL
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.

-30° to -10°C