Please login to your online account to display your discounted pricing

Trilaurin, 98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC421831000

 View more versions of this product

Catalog No. AC421831000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Assay ≥97.5%
Formula Weight 639.01
Quantity 100g
Color White
Density 0.9000g/mL
Chemical Name or Material Trilaurin
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C39H74O6
MDL Number MFCD00026559
CAS 538-24-9
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Trilaurin
CAS 538-24-9
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C39H74O6
Formula Weight 639.01
MDL Number MFCD00026559