missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Trilaurin, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 421830250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Trilaurin | |
0.9000g/mL | |
98% | |
C39H74O6 | |
25g | |
trilaurin, glycerol trilaurate, glyceryl tridodecanoate, glyceryl trilaurate, glycerin trilaurate, lauric acid triglyceride, tridodecanoin, lauric acid triglycerin ester, trilauroylglycerol, laurin, tri | |
VMPHSYLJUKZBJJ-UHFFFAOYSA-N | |
2,3-di(dodecanoyloxy)propyl dodecanoate | |
10851 | |
639.01 |
538-24-9 | |
Authentic | |
Glass bottle | |
MFCD00026559 | |
0.9 | |
Solubility in water: insoluble. Other solubilities: soluble in ethanol,diethyl ether,very soluble in acetone,benzene | |
CCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCC)OC(=O)CCCCCCCCCCC | |
639.01 | |
CHEBI:77389 | |
98% |
Chemical Identifiers
538-24-9 | |
639.01 | |
VMPHSYLJUKZBJJ-UHFFFAOYSA-N | |
10851 | |
2,3-di(dodecanoyloxy)propyl dodecanoate |
C39H74O6 | |
MFCD00026559 | |
trilaurin, glycerol trilaurate, glyceryl tridodecanoate, glyceryl trilaurate, glycerin trilaurate, lauric acid triglyceride, tridodecanoin, lauric acid triglycerin ester, trilauroylglycerol, laurin, tri | |
CHEBI:77389 | |
CCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCC)OC(=O)CCCCCCCCCCC |
Safety and Handling
EINECSNumber : 208-687-
RUO â Research Use Only