missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triisopropylsilane, TCI America™

Manufacturer:  TCI America T1533100ML

 View more versions of this product

Catalog No. T1533100ML



Specifications

Specifications

Triisopropylsilane
Colorless
6485-79-6
MFCD00009657
triisopropylsilane, tris propan-2-yl silane, triisopropyl silane, tri propan-2-yl silicon, triisopropylsilane, tri-isopropylsilyl radical, triisopropylsilyl, tri-iso-propylsilane, ambotzrl-1102, pubchem12855
CC(C)[Si](C(C)C)C(C)C
157.352
158.36
≥98.0% (GC)
166°C
100mL
C9H21Si
1993
ZGYICYBLPGRURT-UHFFFAOYSA-N
tri(propan-2-yl)silicon
6327611
Clear Liquid at 20°C

Chemical Identifiers

6485-79-6
157.352
ZGYICYBLPGRURT-UHFFFAOYSA-N
6327611
CC(C)[Si](C(C)C)C(C)C
C9H21Si
MFCD00009657
triisopropylsilane, tris propan-2-yl silane, triisopropyl silane, tri propan-2-yl silicon, triisopropylsilane, tri-isopropylsilyl radical, triisopropylsilyl, tri-iso-propylsilane, ambotzrl-1102, pubchem12855
tri(propan-2-yl)silicon
Safety and Handling

Safety and Handling

TSCA : No

Documents