Please login to your online account to display your discounted pricing

Triisopropylsilane, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Triisopropylsilane
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C9 H22 Si
Linear Formula [(CH3)2CH]3SiH
Formula Weight 158.36
MDL Number MFCD00009657
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC372410100

  SDS
thermo fisher scientific
AC372410100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Liquid
Formula Weight 158.36
Quantity 10g
Color Clear colorless
Density 0.7730g/mL
Beilstein 04, III, 1851
Chemical Name or Material Triisopropylsilane
Packaging Glass bottle
Refractive Index 1.4330 to 1.4360 (20°C, 589nm)
Assay Percent Range 95%
Molecular Formula C9 H22 Si
MDL Number MFCD00009657
CAS 6485-79-6
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Linear Formula [(CH3)2CH]3SiH
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.

Reduction reagent

Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.