missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triisopropylsilane, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC372410100

 View more versions of this product

Catalog No. AC372410100

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Triisopropylsilane
Clear colorless
6485-79-6
Glass bottle
Liquid
95%
[(CH3)2CH]3SiH
MFCD00009657
0.7730g/mL
10g
Authentic
1.4330 to 1.4360 (20°C, 589nm)
≥94%
C9 H22 Si
158.36
04, III, 1851

Chemical Identifiers

95%
[(CH3)2CH]3SiH
MFCD00009657
≥94%
C9 H22 Si
158.36
Triisopropylsilane

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning