Please login to your online account to display your discounted pricing

Alfa Aesar™ Triisopropylphosphine, 98%

Chemical Identifiers

Chemical Name or Material Triisopropylphosphine
CAS 6476-36-4
Formula Weight 160.24
MDL Number MFCD00015029
UN Number UN2845
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4168402

  SDS
alfa aesar™
4168402
0.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4168409

  SDS
alfa aesar™
4168409
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4168404

  SDS
alfa aesar™
4168404
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Triisopropylphosphine
Density 0.839g/mL
Flash Point −17° (1°F)
Odor Pungent
Sensitivity Air Sensitive
Boiling Point 176° to 178°C
Quantity 0.5g
CAS 6476-36-4
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H250-H252-H314-H318
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.
Refractive Index 1.4670
Physical Form Liquid
Assay 98%
Formula Weight 160.24
MDL Number MFCD00015029
UN Number UN2845
Safety and Handling

Danger

GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H250-H252-H314-H318
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Causes severe skin burns and eye damage.
Causes serious eye damage.