Please login to your online account to display your discounted pricing

Triisopropyl orthoformate, 95%, ACROS Organics™


Quantity 100mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC325441000

 View more versions of this product

Catalog No. AC325441000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 190.29
Assay ≥94%
Physical Form Liquid
Quantity 100mL
Color Clear colorless
Density 0.86g/mL
Packaging Glass bottle
Chemical Name or Material Triisopropyl orthoformate
Linear Formula HC(OCH(CH3))3
Infrared Spectrum Authentic
Assay Percent Range 95%
Molecular Formula C10H22O3
CAS 4447-60-3
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.
MDL Number MFCD00060628
Refractive Index 1.3950 to 1.3980 (20°C, 589nm)
Health Hazard 1 GHS Signal Word: Warning

Specifications

Chemical Name or Material Triisopropyl orthoformate
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C10H22O3
Linear Formula HC(OCH(CH3))3
Formula Weight 190.29
MDL Number MFCD00060628
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.

SDS & Safety Documents