missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Triisopropoxyvanadium(V) Oxide 97.0+%, TCI America™
Supplier: TCI America T25485G
Specifications
Triisopropoxyvanadium(V) Oxide | |
-2°C | |
102°C | |
MFCD00015017 | |
vanadium v oxytriisopropoxide, oxotris propan-2-olato vanadium, vanadium v triisopropoxide oxide, oxotri isopropoxo vanadium, vtip, vo oipr 3, vanadium triisopropoxy oxide, triisopropoxy oxo vanadium, vanadium v tri-isopropoxy oxide, dsggjxauqhkogq-uhfffaoysa-n | |
O=[V].CC(C)O.CC(C)O.CC(C)O | |
247.23 | |
244.20 | |
Liquid |
5588-84-1 | |
Yellow | |
C9H24O4V | |
5 g | |
JOUSPCDMLWUHSO-UHFFFAOYSA-N | |
oxovanadium;propan-2-ol | |
79702 | |
≥97.0% (T) |
Chemical Identifiers
5588-84-1 | |
247.23 | |
JOUSPCDMLWUHSO-UHFFFAOYSA-N | |
79702 | |
O=[V].CC(C)O.CC(C)O.CC(C)O |
C9H24O4V | |
MFCD00015017 | |
vanadium v oxytriisopropoxide, oxotris propan-2-olato vanadium, vanadium v triisopropoxide oxide, oxotri isopropoxo vanadium, vtip, vo oipr 3, vanadium triisopropoxy oxide, triisopropoxy oxo vanadium, vanadium v tri-isopropoxy oxide, dsggjxauqhkogq-uhfffaoysa-n | |
oxovanadium;propan-2-ol |
Safety and Handling
EINECSNumber : (2)-3398
TSCA : Yes