missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triisopropanolamine, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC245761000

 View more versions of this product

Catalog No. AC245761000



Description

Description

Specifications

Specifications

Triisopropanolamine
Authentic
138 mPa.s (60°C)
White to Yellow
100g
98%
N(CH2CH(OH)CH3)3
04,762
1,1',1-nitrilotripropan-2-ol, 2-propanol, 1,1',1-nitrilotris, caswell no. 891, tipa, tri-2-propanolamine, tri-iso-propanolamine, triisopropanolamine, tris 2-hydroxy-1-propyl amine, tris 2-hydroxypropyl amine, tris 2-propanol amine
SLINHMUFWFWBMU-UHFFFAOYSA-N
1-[bis(2-hydroxypropyl)amino]propan-2-ol
24730
Crystalline Low Melting Solid
160°C
Glass bottle
190.0°C (23.0 mmHg)
48.0°C to 52.0°C
122-20-3
C9H21NO3
MFCD00004533
17,371; 12,445; 13,46
Solubility in water: soluble. Other solubilities: soluble in ethanol, diethyl ether
CC(CN(CC(C)O)CC(C)O)O
191.27
191.27
98%

Chemical Identifiers

122-20-3
191.27
SLINHMUFWFWBMU-UHFFFAOYSA-N
24730
CC(CN(CC(C)O)CC(C)O)O
C9H21NO3
MFCD00004533
1,1',1-nitrilotripropan-2-ol, 2-propanol, 1,1',1-nitrilotris, caswell no. 891, tipa, tri-2-propanolamine, tri-iso-propanolamine, triisopropanolamine, tris 2-hydroxy-1-propyl amine, tris 2-hydroxypropyl amine, tris 2-propanol amine
1-[bis(2-hydroxypropyl)amino]propan-2-ol
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.

GHS Signal Word: Warning

EINECSNumber : 204-528-4

SDS Documents