Please login to your online account to display your discounted pricing

Triisobutylaluminium, 1.1M solution in toluene, AcroSeal™, ACROS Organics™


Quantity 100mL
Packaging AcroSeal™ Glass Bottle in Tin Can

Manufacturer: thermo fisher scientific  AC185421000

 View more versions of this product

Catalog No. AC185421000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Solution
Formula Weight 198.33
Quantity 100mL
Color Clear colorless to light yellow
Fieser 09,486
Merck Index 10, 316
Packaging AcroSeal™ Glass Bottle in Tin Can
Assay 24.5 to 25.5% (on Al basis)
Density 0.8400g/mL
CAS 108-88-3
Linear Formula [(CH3)2CHCH2]3Al
Synonym TIBAL
Health Hazard 2 GHS H Statement:
May be fatal if swallowed and enters airways.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
May cause damage to organs through prolonged or repeated exposure.
Suspected of damaging the unborn child.
Highly flammable liquid an
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Store in a dry place. Store in a closed container.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Danger
Name Note 1.1M Solution in Toluene
Chemical Name or Material Triisobutylaluminium
CAS 100-99-2
Molecular Formula C12H27Al
MDL Number MFCD00008929

Specifications

Chemical Name or Material Triisobutylaluminium
Name Note 1.1M Solution in Toluene
CAS 100-99-2
CAS 108-88-3
Assay 24.5 to 25.5% (on Al basis)
Molecular Formula C12H27Al
Linear Formula [(CH3)2CHCH2]3Al
Formula Weight 198.33
MDL Number MFCD00008929
Synonym TIBAL
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Store in a dry place. Store in a closed container.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May be fatal if swallowed and enters airways.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
May cause damage to organs through prolonged or repeated exposure.
Suspected of damaging the unborn child.
Highly flammable liquid an

SDS & Safety Documents