Please login to your online account to display your discounted pricing

Triiron dodecacarbonyl, 99%, stabilized, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC209290250

 View more versions of this product

Catalog No. AC209290250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder or Crystals
Formula Weight 503.66
Quantity 25g
Color Black
Density 1.9900g/mL
Chemical Name or Material Triiron dodecacarbonyl
Packaging Glass bottle
Stabilizer 5 to 10% MeOH
Assay Percent Range 99%
Molecular Formula C12Fe3O12
Linear Formula Fe3(CO)12
MDL Number MFCD00135617
CAS 17685-52-8
Health Hazard 1 Warning
Health Hazard 2 GHS P Statement Flammable solid. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. May cause damage to organs.
Health Hazard 3 GHS P Statement Keep container tightly closed. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection.
Infrared Spectrum Authentic
Elemental Analysis 98.5% min.

Specifications

Chemical Name or Material Triiron dodecacarbonyl
Assay Percent Range 99%
Molecular Formula C12Fe3O12
Linear Formula Fe3(CO)12
Formula Weight 503.66
MDL Number MFCD00135617
Accessories

Warning

GHS P Statement Keep container tightly closed. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection.

GHS P Statement Flammable solid. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. May cause damage to organs.

SDS & Safety Documents